Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029555.2 | NP_083831.1 | 7 | 676 | ||
GenBank mRNA | AK002661.1 | - | 7 | 674 | ||
AY279096.1 | - | 7 | 589 | |||
BC038151.1 | - | 7 | 603 | |||
BC051507.1 | - | 7 | 717 | |||
BC058163.1 | - | 7 | 609 | 5100572 |