Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029561.4 | NP_083837.2 | 8 | 1088 | ||
GenBank mRNA | AK002978.1 | - | 6 | 607 | ||
AK031073.1 | - | 8 | 903 | |||
AK049678.1 | - | 8 | 958 | |||
AK078939.1 | - | 8 | 998 | |||
AK132235.1 | - | 8 | 1017 | |||
AK150832.1 | - | 8 | 902 | |||
BC071237.1 | - | 8 | 1007 | 5003738 |