Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281463.1 | NP_001268392.1 | 26 | 5268 | ||
NM_006306.3 | NP_006297.2 | 25 | 5122 | |||
GenBank mRNA | D80000.2 | - | 25 | 4994 | ||
S78271.1 | - | 25 | 5025 |