Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029911.4 | NP_084187.2 | 2, 3 | 498 | ||
GenBank mRNA | AK006295.1 | - | 3, 4 | 722 | ||
AK019376.1 | - | 2, 3 | 901 | |||
AK031904.1 | - | 2, 3 | 429 | |||
AK036066.1 | - | 2, 3 | 772 | |||
AK082153.1 | - | 2, 3 | 499 | |||
BC132487.1 | - | 2, 3 | 440 | 40130764 | ||
BC137869.1 | - | 2, 3 | 440 | 8860888 | ||
DQ185134.1 | - | 2, 3 | 395 |