Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rai14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166408.1 | NP_001159880.1 | 9 | 793 | ||
NM_030690.3 | NP_109615.1 | 9 | 879 | |||
GenBank mRNA | AF202315.1 | - | 8 | 666 | ||
AF274866.1 | - | 9 | 858 | |||
AK129333.1 | - | 9 | 729 | |||
AK141233.1 | - | 5 | 379 | |||
AK160119.1 | - | 9 | 789 | |||
BC052458.1 | - | 9 | 879 | 6830677 |