Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033371.2 | NP_203535.1 | 11 | 2038 | ||
GenBank mRNA | AK013817.1 | - | 4 | 1032 | ||
AK134809.1 | - | 3 | 1004 | |||
AK150235.1 | - | 8 | 833 | |||
AK154865.1 | - | 11 | 2023 | |||
AK171452.1 | - | 11 | 2023 | |||
AK191582.1 | - | 1 | 38 | |||
AK220248.1 | - | 9 | 1836 | |||
AY010723.1 | - | 10 | 1233 | |||
BC033318.1 | - | 8 | 1396 | |||
BC044775.1 | - | 9 | 1793 | |||
BC049958.1 | - | 9 | 1793 | |||
BC057450.1 | - | 10 | 1655 | 3966334 | ||
BC096620.1 | - | 11 | 2038 | 6390076 |