Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080420.2 | NP_536345.2 | 4 | 568 | ||
GenBank mRNA | BC016212.1 | - | 4 | 575 | 4924899 |