Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080637.3 | NP_542368.2 | 3, 4 | 454 | ||
GenBank mRNA | AF343565.1 | - | 2, 3 | 343 | ||
AK131722.1 | - | 3, 4 | 455 |