Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159581.1 | NP_001153053.1 | 4 | 363 | ||
NM_133250.2 | NP_573513.2 | 3 | 270 | |||
GenBank mRNA | AB117937.1 | - | 2 | 232 | ||
AB117938.1 | - | 2 | 232 | |||
AB117939.1 | - | 4 | 347 | |||
AK017730.1 | - | 3 | 258 | |||
AK077546.1 | - | 3 | 267 | |||
AK156790.1 | - | 4 | 353 | |||
AY007717.1 | - | 2 | 226 | |||
BC057942.1 | - | 3 | 251 | 5320744 | ||
CX213088.1 | - | 3 | 305 |