Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133346.2 | NP_579924.1 | 6 | 1618 | ||
GenBank mRNA | AF398967.1 | - | 3656 | |||
AK010909.1 | - | 6 | 1599 | |||
AK049312.1 | - | 6 | 1607 | |||
AK088890.1 | - | 6 | 1553 | |||
BC019192.1 | - | 6 | 1610 | 5035395 | ||
BC085485.1 | - | 5 | 2299 |