Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133733.4 | NP_598494.2 | 2, 3 | 548 | ||
GenBank mRNA | AB040490.1 | - | 2, 3 | 465 | ||
AK028983.1 | - | 2, 3 | 518 | |||
AK033723.1 | - | 2, 3 | 550 | |||
AK080481.1 | - | 2, 3 | 503 | |||
AY259213.1 | - | 2, 4 | 177 | |||
AY326421.1 | - | 2, 3 | 183 | |||
BC026447.1 | - | 2, 3 | 374 | 4459717 |