Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133739.2 | NP_598500.1 | 3, 4 | 560 | ||
GenBank mRNA | AK010163.1 | - | 1, 2 | 53 | ||
AK150361.1 | - | 3, 4 | 508 | |||
AK150365.1 | - | 4, 5 | 588 | |||
AK151044.1 | - | 3, 4 | 528 | |||
AK152180.1 | - | 3, 4 | 508 | |||
AK152423.1 | - | 3, 4 | 473 | |||
AK153122.1 | - | 3, 4 | 473 | |||
AK153378.1 | - | 4, 5 | 588 | |||
AK153539.1 | - | 3, 4 | 508 | |||
AK155767.1 | - | 3, 4 | 528 | |||
AK157001.1 | - | 4, 5 | 485 | |||
AK162610.1 | - | 3, 4 | 556 | |||
AK165671.1 | - | 3, 4 | 527 | |||
AK166444.1 | - | 3, 4 | 473 | |||
AK170823.1 | - | 3, 4 | 519 | |||
AK172352.1 | - | 3, 4 | 489 | |||
AK172553.1 | - | 3, 4 | 489 | |||
AK172701.1 | - | 3, 4 | 489 | |||
BC010292.1 | - | 3, 4 | 509 | 3153174 | ||
BC086319.1 | - | 3, 4 | 517 | 5293409 |