Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133747.2 | NP_598508.2 | 4 | 603 | ||
GenBank mRNA | AK012997.1 | - | 4 | 537 | ||
AK046481.1 | - | 4 | 520 | |||
AK146215.1 | - | 4 | 547 | |||
AK146761.1 | - | 4 | 569 | |||
AK146955.1 | - | 4 | 516 | |||
AK167224.1 | - | 4 | 487 | |||
BC004626.1 | - | 4 | 500 | 3585698 |