Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163833.1 | NP_001157305.1 | 2 | 1816 | ||
GenBank mRNA | AK006842.1 | - | 1 | 1217 | ||
AK044179.1 | - | 1 | 1216 | |||
AK155382.1 | - | 2 | 1802 | |||
AK172141.1 | - | 198 | ||||
BC013191.1 | - | 1 | 1418 | |||
BC037487.1 | - | 2 | 1827 |