Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133756.4 | NP_598517.1 | 14 | 1387 | ||
GenBank mRNA | AK010393.1 | - | 14 | 1400 | ||
AK077645.1 | - | 14 | 1388 | |||
AK145957.1 | - | 1384 | ||||
BC020174.1 | - | 14 | 1387 | 4505118 | ||
BC096466.1 | - | 14 | 1380 | 3711417 |