Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133762.3 | NP_598523.3 | 19 | 2375 | ||
GenBank mRNA | AY455829.1 | - | 13 | 1486 | ||
BC008539.1 | - | 8 | 1056 | |||
BC076631.1 | - | 18 | 2185 | 30637859 | ||
CK636577.1 | - | 3 | 196 |