Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133762.3 | NP_598523.3 | 29 | 4483 | ||
GenBank mRNA | AK049215.1 | - | 1 | 725 | ||
AK139749.1 | - | 2 | 1033 | |||
AY455829.1 | - | 23 | 3598 | |||
BC008539.1 | - | 18 | 3163 | |||
BC076631.1 | - | 28 | 4293 | 30637859 |