Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006324.2 | NP_006315.1 | 4 | 646 | ||
XM_011522814.1 | XP_011521116.1 | 4 | 669 | |||
XM_011522815.1 | XP_011521117.1 | 4 | 669 | |||
GenBank mRNA | AB009285.1 | - | 4 | 618 | ||
AK226031.1 | - | 5 | 634 | |||
AK313948.1 | - | 4 | 588 | |||
BC000991.2 | - | 4 | 642 | 3449836 | ||
BT009819.1 | - | 4 | 509 | |||
CR542111.1 | - | 4 | 509 | |||
D85939.1 | - | 4 | 618 | |||
D86549.1 | - | 3 | 445 | |||
LC057679.1 | - | 4 | 509 | |||
LC057680.1 | - | 3 | 391 | |||
LC057682.1 | - | 4 | 509 | |||
LC057683.1 | - | 4 | 509 | |||
LC057684.1 | - | 3 | 391 |