Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133858.4 | NP_598619.2 | 2 | 1185 | ||
NM_199475.1 | NP_955769.1 | 2 | 1185 | |||
GenBank mRNA | AB075603.1 | - | 1 | 196 | ||
AK036838.1 | - | 2 | 1177 | |||
AK087859.1 | - | 2 | 998 | |||
AK162551.1 | - | 2 | 905 | |||
AK191590.1 | - | 1 | 372 | |||
AK211364.1 | - | 1 | 338 | |||
BC051048.1 | - | 1 | 68 | |||
BC094388.1 | - | 2 | 1002 | 5708548 | ||
BC132301.1 | - | 1 | 430 | 40130578 | ||
BC144749.1 | - | 1 | 430 | 9053297 |