Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133858.4 | NP_598619.2 | 2 | 1332 | ||
NM_199475.1 | NP_955769.1 | 2 | 1332 | |||
GenBank mRNA | AB075603.1 | - | 1 | 343 | ||
AK036838.1 | - | 2 | 1324 | |||
AK087859.1 | - | 2 | 1145 | |||
AK162551.1 | - | 2 | 1052 | |||
AK169673.1 | - | 2 | 1332 | |||
BC039762.1 | - | 1 | 150 | |||
BC051048.1 | - | 1 | 215 | |||
BC094388.1 | - | 2 | 1149 | 5708548 | ||
BC132301.1 | - | 1 | 577 | 40130578 | ||
BC144749.1 | - | 1 | 577 | 9053297 |