Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205157.1 | NP_001192086.1 | 9 | 1144 | ||
NM_138303.2 | NP_612176.1 | 8 | 1282 | |||
GenBank mRNA | AK033705.1 | - | 9 | 1093 | ||
AK133631.1 | - | 9 | 1104 | |||
BC002282.1 | - | 8 | 1197 | 3497525 |