Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139140.1 | NP_631879.1 | 13 | 2004 | ||
GenBank mRNA | AB079132.1 | - | 13 | 2004 | ||
AK012237.1 | - | 2 | 385 | |||
AK159335.1 | - | 13 | 2099 | |||
AK167788.1 | - | 12 | 1682 | |||
AK171266.1 | - | 12 | 1760 | |||
BC023432.1 | - | 11 | 1654 | 5029821 | ||
BC072579.1 | - | 13 | 2271 |