Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144539.5 | NP_653122.2 | 4 | 848 | ||
GenBank mRNA | AB196816.1 | - | 3 | 395 | ||
AB196817.1 | - | 3 | 395 | |||
AB196818.1 | - | 3 | 395 | |||
AB196819.1 | - | 3 | 395 | |||
AB196837.1 | - | 3 | 395 | |||
AF467909.1 | - | 3 | 395 | |||
AF467910.1 | - | 3 | 395 | |||
AF467911.1 | - | 3 | 395 | |||
AK016183.1 | - | 4 | 652 | |||
AK030135.1 | - | 4 | 709 | |||
AK030148.1 | - | 4 | 849 | |||
AK040678.1 | - | 3 | 476 | |||
AK089525.1 | - | 3 | 476 | |||
AK172665.1 | - | 3 | 442 | |||
BC011154.1 | - | 3 | 407 | 4168309 |