Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144802.4 | NP_659051.3 | 4 | 1019 | ||
GenBank mRNA | AK012866.1 | - | 4 | 735 | ||
AK034860.1 | - | 4 | 964 | |||
AK048900.1 | - | 4 | 808 | |||
AK146879.1 | - | 4 | 981 | |||
AK160943.1 | - | 3 | 827 | |||
AK171091.1 | - | 4 | 886 | |||
BC012849.1 | - | 4 | 965 | 3587716 |