Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144802.4 | NP_659051.3 | 4, 5 | 1053 | ||
GenBank mRNA | AK012866.1 | - | 4, 5 | 769 | ||
AK034860.1 | - | 4, 5 | 998 | |||
AK048900.1 | - | 4, 5 | 842 | |||
AK146879.1 | - | 4, 5 | 1015 | |||
AK160943.1 | - | 3, 4 | 861 | |||
AK171091.1 | - | 4, 5 | 920 | |||
BC004763.1 | - | 1, 2 | 26 | |||
BC012849.1 | - | 4, 5 | 999 | 3587716 |