Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145221.2 | NP_660256.1 | 22 | 2359 | ||
GenBank mRNA | AK030445.1 | - | 22 | 2238 | ||
AK033566.1 | - | 22 | 2330 | |||
AK045438.1 | - | 22 | 2295 | |||
AK146356.1 | - | 22 | 2330 | |||
AK173169.1 | - | 7 | 810 | |||
AY113705.1 | - | 22 | 2324 | |||
BC019708.1 | - | 14 | 1637 | |||
BC063751.1 | - | 22 | 2371 | 6511401 |