Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145823.2 | NP_665822.1 | 6 | 1172 | ||
GenBank mRNA | AB077281.1 | - | 6 | 1153 | ||
AB077282.1 | - | 6 | 450 | |||
AK003844.1 | - | 5 | 420 | |||
AK021206.1 | - | 6 | 385 | |||
AK153625.1 | - | 6 | 1173 | |||
AK171340.1 | - | 6 | 1199 | |||
AK171918.1 | - | 6 | 1169 | |||
BC082333.1 | - | 6 | 1004 | |||
BC108351.1 | - | 6 | 1032 | 6312193 |