Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152814.2 | NP_690027.1 | 4 | 605 | ||
GenBank mRNA | AK012367.1 | - | 4 | 583 | ||
BC019790.1 | - | 4 | 492 | 5151196 |