Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152822.3 | NP_690035.2 | 1, 2 | 322 | ||
GenBank mRNA | AK018059.1 | - | 1, 2 | 252 | ||
AK089160.2 | - | 1, 2 | 274 | |||
AK163919.1 | - | 1, 2 | 311 | |||
BC032218.1 | - | 1, 2 | 245 | |||
BC141155.1 | - | 1, 2 | 259 | 9055767 | ||
BY036255.1 | - | 1, 2 | 287 | |||
BY059480.1 | - | 1, 2 | 323 |