Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152822.3 | NP_690035.2 | 5 | 729 | ||
GenBank mRNA | AK018059.1 | - | 5 | 659 | ||
AK089160.2 | - | 4 | 2520 | |||
AK163919.1 | - | 4 | 2556 | |||
BC004735.1 | - | 4 | 321 | |||
BC032218.1 | - | 5 | 652 | |||
BC141155.1 | - | 5 | 666 | 9055767 | ||
BQ573371.1 | - | 4 | 309 |