Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145826.1 | NP_001139298.1 | 8 | 2548 | ||
NM_153406.3 | NP_700455.3 | 5 | 2319 | |||
GenBank mRNA | AB093236.1 | - | 8 | 2548 | ||
AK035500.1 | - | 5 | 2320 | |||
AK041543.1 | - | 2 | 812 | |||
AY884297.1 | - | 5 | 2320 | |||
BC037464.1 | - | 2 | 1406 | 4194868 | ||
BC072595.1 | - | 4 | 2158 | 6408832 |