Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006351.3 | NP_006342.2 | 3, 4 | 485 | ||
GenBank mRNA | AF026030.1 | - | 3, 4 | 302 | ||
AF041254.1 | - | 3, 4 | 343 | |||
AK223062.1 | - | 3, 4 | 328 | |||
AK289634.1 | - | 3, 4 | 485 | |||
BC033628.1 | - | 3, 4 | 315 | 5248163 | ||
X58234.1 | - | 23 |