Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn2aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172407.3 | NP_765995.1 | 2 | 533 | ||
GenBank mRNA | AK014874.1 | - | 1 | 79 | ||
AK048437.1 | - | 2 | 502 | |||
AK133831.1 | - | 2 | 533 |