Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172413.2 | NP_766001.1 | 3 | 1094 | ||
GenBank mRNA | AK008416.1 | - | 1 | 141 | ||
AK087971.1 | - | 3 | 1041 | |||
AK146057.1 | - | 3 | 1073 | |||
AK146956.1 | - | 3 | 1136 | |||
AK147015.1 | - | 3 | 1136 | |||
BC027363.1 | - | 3 | 1058 | 4951566 | ||
BC050056.1 | - | 3 | 1056 | 30016070 | ||
BC064814.1 | - | 3 | 1090 | 6513822 | ||
BC094890.1 | - | 3 | 1094 | 30649398 |