Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270988.1 | NP_001257917.1 | 5, 6 | 811 | ||
NM_172421.5 | NP_766009.2 | 5, 6 | 811 | |||
GenBank mRNA | AK016141.1 | - | 5, 6 | 517 | ||
AK036839.1 | - | 5, 6 | 768 | |||
AK080679.1 | - | 5, 6 | 785 | |||
AK148015.1 | - | 5, 6 | 816 | |||
BC141140.1 | - | 5, 6 | 645 | 9055752 |