Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172597.2 | NP_766185.1 | 5 | 610 | ||
GenBank mRNA | AK017582.1 | - | 5 | 505 | ||
AK034025.1 | - | 5 | 599 | |||
AK039945.1 | - | 5 | 477 | |||
AK046466.1 | - | 5 | 611 | |||
AK173153.1 | - | 1107 | ||||
AK215424.1 | - | 3 | 190 | |||
BC052488.1 | - | 5 | 486 | 6438726 |