Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172597.2 | NP_766185.1 | 5, 6 | 623 | ||
GenBank mRNA | AK017582.1 | - | 5, 6 | 518 | ||
AK034025.1 | - | 5, 6 | 612 | |||
AK039945.1 | - | 5, 6 | 490 | |||
AK046466.1 | - | 5, 6 | 624 | |||
AK173153.1 | - | 1120 | ||||
AK215424.1 | - | 3, 4 | 203 | |||
BC052488.1 | - | 5, 6 | 499 | 6438726 |