Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006360.5 | NP_006351.2 | 2, 3 | 294 | ||
XM_011519840.2 | XP_011518142.1 | 2, 3 | 318 | |||
GenBank mRNA | AF064603.1 | - | 2, 3 | 224 | ||
AK131064.1 | - | 2, 3 | 248 | |||
AK222564.1 | - | 2, 3 | 213 | |||
AK292139.1 | - | 2, 3 | 236 | |||
AK300002.1 | - | 2, 3 | 238 | |||
AK312512.1 | - | 2, 3 | 263 | |||
AY769947.1 | - | 2, 3 | 173 | |||
BC019103.2 | - | 2, 3 | 216 | 5021401 | ||
BC051292.1 | - | 2, 3 | 218 | 3607828 | ||
CR450300.1 | - | 2, 3 | 173 | |||
DB479224.1 | - | 2, 3 | 294 | |||
DQ185042.1 | - | 2, 3 | 164 |