Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iws1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173441.3 | NP_775617.1 | 3 | 1273 | ||
GenBank mRNA | AK028247.1 | - | 3 | 1244 | ||
AK033013.1 | - | 3 | 1269 | |||
BC068184.1 | - | 3 | 1213 | 30639737 |