Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114311.1 | NP_001107783.1 | 3 | 2148 | ||
NM_001286376.1 | NP_001273305.1 | 3 | 2148 | |||
NM_175162.4 | NP_780371.3 | 3 | 943 | |||
GenBank mRNA | AK030015.1 | - | 3 | 757 | ||
AK053894.1 | - | 3 | 2146 | |||
AK129346.1 | - | 3 | 2108 | |||
AK162847.1 | - | 3 | 977 | |||
BC099949.1 | - | 3 | 957 | 30096824 | ||
BC100572.1 | - | 3 | 943 | 30085660 | ||
BC100573.1 | - | 3 | 943 | 30086796 | ||
BC138237.1 | - | 3 | 714 | 8861256 | ||
BC138238.1 | - | 3 | 714 | 8861257 | ||
BC145211.1 | - | 4 | 816 |