Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083810.2 | NP_001077279.1 | 6 | 833 | ||
NM_001110849.1 | NP_001104319.1 | 7 | 696 | |||
NM_175181.5 | NP_780390.3 | 6 | 782 | |||
GenBank mRNA | AK011178.1 | - | 1 | 30 | ||
AK030499.1 | - | 6 | 741 | |||
AK145981.1 | - | 6 | 784 | |||
BC048394.1 | - | 6 | 692 | 6391924 | ||
BC172132.1 | - | 5 | 508 | 9054802 | ||
BC172133.1 | - | 5 | 508 |