Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006369.4 | NP_006360.3 | 6 | 1942 | ||
XM_017000074.1 | XP_016855563.1 | 6 | 1968 | |||
GenBank mRNA | AF370430.1 | - | 6 | 1842 | ||
AK125827.1 | - | 5 | 3219 | |||
AK223420.1 | - | 3 | 1304 | |||
AK291283.1 | - | 6 | 1925 | |||
AK296436.1 | - | 6 | 1761 | |||
AK297596.1 | - | 6 | 1956 | |||
BC004953.1 | - | 3 | 1178 | |||
BC101522.1 | - | 6 | 1932 | 8069028 | ||
BC101524.1 | - | 6 | 1932 | 8069030 | ||
BX537366.1 | - | 6 | 1852 | |||
X86018.1 | - | 3 | 1154 |