Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293784.1 | NP_001280713.1 | 8, 9 | 1034 | ||
NM_175188.4 | NP_780397.2 | 7, 8 | 981 | |||
GenBank mRNA | AK036489.1 | - | 3, 4 | 545 | ||
AK086052.1 | - | 8, 9 | 993 | |||
AK152592.1 | - | 3, 4 | 546 | |||
BC066008.1 | - | 7, 8 | 981 | 5703753 |