Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hepacam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175189.4 | NP_780398.2 | 5, 6 | 946 | ||
GenBank mRNA | AK013637.1 | - | 5, 6 | 946 | ||
AK081154.1 | - | 5, 6 | 862 | |||
AK138438.1 | - | 4, 5 | 470 | |||
AK158265.1 | - | 5, 6 | 1184 | |||
BC082537.1 | - | 5, 6 | 860 | |||
BC139057.1 | - | 5, 6 | 936 | 8862079 | ||
BC139058.1 | - | 5, 6 | 936 | 8862080 |