Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006373.3 | NP_006364.2 | 6 | 1872 | ||
GenBank mRNA | AK097072.1 | - | 7 | 1699 | ||
BC001913.1 | - | 6 | 1741 | |||
BC008725.2 | - | 6 | 1796 | 3162543 | ||
BC014279.2 | - | 6 | 1778 | 3834206 | ||
BC015041.2 | - | 6 | 1810 | 3919901 | ||
BC090858.1 | - | 6 | 1810 | 6528247 | ||
U18009.1 | - | 6 | 1498 | |||
U25779.1 | - | 6 | 1480 |