Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038993.3 | NP_001034082.1 | 4 | 493 | ||
NM_175201.5 | NP_780410.2 | 3 | 523 | |||
GenBank mRNA | AK006888.1 | - | 3 | 485 | ||
AK044324.1 | - | 3 | 338 | |||
AK147608.1 | - | 4 | 550 | |||
BC060730.1 | - | 3 | 500 | 6408623 | ||
BC062976.1 | - | 3 | 437 | 6848803 |