Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 13 | 1627 | ||
GenBank mRNA | AK018051.1 | - | 12 | 1316 | ||
AK035336.1 | - | 8 | 909 | |||
AK038285.1 | - | 12 | 1375 | |||
BC141358.1 | - | 13 | 1622 | 9055970 |