Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 17 | 2047 | ||
GenBank mRNA | AK038285.1 | - | 16 | 1795 | ||
BC141358.1 | - | 17 | 2042 | 9055970 |