Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYOU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130991.2 | NP_001124463.1 | 17 | 2062 | ||
NM_006389.4 | NP_006380.1 | 17 | 2195 | |||
XM_005271392.3 | XP_005271449.1 | 17 | 2056 | |||
XM_005271393.3 | XP_005271450.1 | 17 | 2036 | |||
XM_005271394.3 | XP_005271451.1 | 17 | 2103 | |||
XM_017017095.1 | XP_016872584.1 | 17 | 2091 | |||
XM_017017096.1 | XP_016872585.1 | 17 | 2112 | |||
XM_017017097.1 | XP_016872586.1 | 17 | 2035 | |||
GenBank mRNA | AK127324.1 | - | 16 | 2064 | ||
AK222756.1 | - | 17 | 2088 | |||
AK299648.1 | - | 10 | 986 | |||
AK301514.1 | - | 15 | 1754 | |||
AK304264.1 | - | 16 | 2161 | |||
AK314178.1 | - | 17 | 2086 | |||
BC072436.1 | - | 17 | 2061 | 5756154 | ||
DQ350134.1 | - | 17 | 2083 | |||
DQ372932.1 | - | 17 | 2083 | |||
U65785.1 | - | 17 | 2063 |