Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176860.5 | NP_789830.1 | 5 | 777 | ||
GenBank mRNA | AB075602.1 | - | 5 | 772 | ||
AK013361.1 | - | 5 | 845 | |||
AK034450.1 | - | 5 | 968 | |||
AK035764.1 | - | 5 | 778 | |||
AK133948.1 | - | 5 | 775 | |||
AK150909.1 | - | 5 | 1344 | |||
AK151721.1 | - | 5 | 1345 | |||
AK151895.1 | - | 5 | 978 | |||
AK152013.1 | - | 5 | 978 | |||
AK154576.1 | - | 5 | 999 | |||
AK172288.1 | - | 778 | ||||
BC053436.1 | - | 5 | 749 | 6510702 |